MilliporeSigma
[Study on GC fingerprint of the constituents in Herba Asari].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-02-15)
Feng Zhang, Shao-ping Fu, Qing Xu, Hong-bin Xiao, Shao-qing Cai, Xin-miao Liang
ABSTRACT
To establish a method for GC fingerprint determination of the chemical constituents in Herba Asari. GC and GC-MS were used to optimize the fingerprint determination method, and identify the main peaks in the GC fingerprint. A preferable method for GC fingerprint determination of the chemical constituents in Herba Asari was established. A general acquaintance of the chemical constituents in Herba Asari can be obtained by using the preferable GC fingerprint determination method, which is useful for quality evaluation of the crude drug of Herba Asari.
Sigma-Aldrich
3,5-Dimethoxytoluene, 98%