MilliporeSigma
  • Chain-length-dependent helical motifs and self-association of beta-peptides with constrained side chains.
Chain-length-dependent helical motifs and self-association of beta-peptides with constrained side chains.
Journal of the American Chemical Society (2005-01-13)
Anasztázia Hetényi, István M Mándity, Tamás A Martinek, Gábor K Tóth, Ferenc Fülöp
ABSTRACT
Homo-oligomers constructed by using trans-2-aminocyclohexanecarboxylic acid monomers without protecting groups were studied. Both ab initio theory and NMR measurements showed that the tetramer tends to adopt a 10-helix motif, while the pentamer and hexamer form the known 14-helix. It was concluded that the conformationally constrained backbone is flexible enough to afford both 10-helical and 14-helical motifs, this observation in turn providing evidence of the true folding process. Self-association of the helical units was also detected, and the results of variable-temperature diffusion NMR measurements strongly suggested the presence of helical bundles in methanol solution.