MilliporeSigma
  • Enantio-selective inhibition of (1R,9S)- and (1S,9R)-beta-hydrastines on dopamine biosynthesis in PC12 cells.
Enantio-selective inhibition of (1R,9S)- and (1S,9R)-beta-hydrastines on dopamine biosynthesis in PC12 cells.
Neuropharmacology (2004-11-24)
Shou Yu Yin, Yu Mi Kim, Jae Joon Lee, Chun Mei Jin, Yoo Jung Yang, Jing Jie Ma, Min Hee Kang, Masaaki Kai, Myung Koo Lee
ABSTRACT
The inhibitory effects of (1R,9S)- and (1S,9R)-enantiomers of beta-hydrastine (BHS) on dopamine biosynthesis in PC12 cells were investigated. (1R,9S)-BHS decreased the intracellular dopamine content with the IC50 value of 14.3 microM at 24 h, but (1S,9R)-BHS did not. (1R,9S)-BHS was not cytotoxic at concentrations up to 250 microM towards PC12 cells. In these conditions, (1R,9S)-BHS inhibited tyrosine hydroxylase (TH) activity mainly in a concentration-dependent manner (33% inhibition at 20 microM) and decreased TH mRNA level in PC12 cells. The inhibitory patterns of dopamine content and TH activity by (1R,9S)-BHS showed similar behavioral curves. (1R,9S)-BHS at 10-50 microM also reduced the intracellular cyclic AMP level and Ca2+ concentration. In addition, treatment of L-DOPA at 20-50 microM for 24 h increased the intracellular dopamine content to 198-251% compared with the control in PC12 cells. However, the increase in dopamine levels induced by L-DOPA (20-50 microM) was reduced when L-DOPA was combined with (1R,9S)-BHS (10-50 microM). These results indicate that (1R,9S)-BHS, but not (1S,9R)-BHS, reduced dopamine content and L-DOPA-induced increase in dopamine content, in part, through the inhibition of TH activity and TH gene expression in PC12 cells: thus, (1R,9S)-BHS proved to have a function to regulate dopamine biosynthesis.
Product Description
Hydrastine hydrochloride, European Pharmacopoeia (EP) Reference Standard