MilliporeSigma
  • [Synthesis and cytotoxic properties of anomeric 5-substituted 3'-azido-2',3'-dideoxyuridines].
[Synthesis and cytotoxic properties of anomeric 5-substituted 3'-azido-2',3'-dideoxyuridines].
Bioorganicheskaia khimiia (1992-03-01)
A A Bakhmedova, I V Iartseva, O S Zhukova, T P Volosiuk, A M Iurkevich, S Ia Mel'nik
ABSTRACT
Glycosylation of trimethylsilyl derivatives of 5-benzyloxymethyl- and 5-hydroxymethyluracil with 3-azido-2,3-dideoxy-5-O-benzoyl-D-ribofuranosyl chloride (prepared from ethyl 3-azido-2,3-dideoxy-5-O-benzoyl-D-ribofuranoside) and subsequent deacylation gave in both cases a mixture of anomeric 3'-azido-2',3'-dideoxy-5-benzyloxymethyl-or 5-hydroxymethyluridines. The anomers were separated by preparative TLC and their structures were studied by UV, IR and 1H-NMR spectroscopy. It is shown that 1-(3-azido-2,3-dideoxy-alpha-D-ribofuranosyl)-5-benzyloxymethyluracil has cytotoxic activity in vitro: in 10(-5)-10(-4) M concentrations it inhibits the thymidine incorporation into DNA of CaOv cells on 78.6-95.2%.