MilliporeSigma
  • Comparison of tropane alkaloid spectra between Datura innoxia grown in Egypt and Bulgaria.
Comparison of tropane alkaloid spectra between Datura innoxia grown in Egypt and Bulgaria.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2004-07-10)
Strahil Berkov, Rawia Zayed
ABSTRACT
The alkaloid spectra of Datura innoxia plants grown in Egypt and Bulgaria were investigated by GC-MS. Thirty-eight alkaloids were detected in the roots, leaves and fruits of the plants. Five new alkaloids for D. innoxia are reported. Alkaloid spectra of Egyptian and Bulgarian plants differ significantly in respect to their alkaloid composition and main alkaloids accumulated in the plant organs.
Sigma-Aldrich
Tropone, 97%