MilliporeSigma
  • Performance of gel and paste bait products for German cockroach (Dictyoptera: Blattellidae) control: laboratory and field studies.
Performance of gel and paste bait products for German cockroach (Dictyoptera: Blattellidae) control: laboratory and field studies.
Journal of economic entomology (1992-08-01)
A G Appel
ABSTRACT
Gel and paste bait formulations containing boric acid or hydramethylnon were evaluated in field and laboratory experiments with the German cockroach, Blattella germanica (L.). Bait formulations contained 14-62% water by weight and lost the most water after 1 d of desiccation. Bait age and water content did not alter the toxicity (LT50) of boric acid-based baits. Toxicity of hydramethylnon (Maxforce) bait, however, increased with bait age. In continuous exposure tests without harborage, boric acid formulations had LT50s of 8.5 and 10.1 d for Blue Diamond and IT WORKS, respectively. Maxforce bait had an LT50 of 4.2 d. LT50s determined with cardboard harborage or in Ebeling choice boxes were 1.5 to 2.2 times lower than those from continuous exposure tests without harborage. Relative repellency, measured as the mean percent of live cockroaches in the light side of the choice box, was greatest for IT WORKS (21.4%) and Maxforce (25.4%) formulations, followed by Blue Diamond (10.9%). A performance index that combined choice box repellency and mortality data indicated that Maxforce had the greatest potential for field effectiveness. Blue Diamond and Maxforce significantly reduced German cockroach populations in the field. Maxforce performed best in field trials when applied at 10 or 20 placements per apartment. Apartments treated with IT WORKS had significantly greater cockroach trap catches. Factors that may affect the predictive power of the performance index are discussed.
Product Description
Supelco
Hydramethylnon, PESTANAL®, analytical standard