MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Determination of triforine using high-performance liquid chromatography with tandem mass spectrometry.
Determination of triforine using high-performance liquid chromatography with tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2004-03-31)
Julie A Harrold, Mark D Sykes, Martin Goodrum, Richard J Fussell
ABSTRACT
A method for determination of triforine using high-performance liquid chromatography with electrospray tandem mass spectrometry was developed. A simple ethyl acetate extraction with solvent exchange into water/methanol was used for sample preparation. The method was validated at 0.01 and 0.05 mg kg(-1) levels in apple, tomato and tinned blackcurrants. Recoveries were in the range 56.6-99.8% and no matrix suppression or enhancement effects were observed.
Product Description
Supelco
Triforin, PESTANAL®, analytical standard