MilliporeSigma
  • A Biacore biosensor method for detailed kinetic binding analysis of small molecule inhibitors of p38alpha mitogen-activated protein kinase.
A Biacore biosensor method for detailed kinetic binding analysis of small molecule inhibitors of p38alpha mitogen-activated protein kinase.
Analytical biochemistry (2004-01-13)
David Casper, Marina Bukhtiyarova, Eric B Springman
ABSTRACT
Protein kinases are emerging as one of the most intensely studied classes of enzymes as their central roles in physiologically and clinically important cellular signaling events become more clearly understood. We report here the development of a real-time, label-free method to study protein kinase inhibitor binding kinetics using surface plasmon resonance-based biomolecular interaction analysis (Biacore). Utilizing p38alpha mitogen-activated protein kinase as a model system, we studied the binding properties of two known small molecule p38alpha inhibitors (SB-203580 and SKF-86002). Direct coupling of p38alpha to the biosensor surface in the presence of a reversible structure-stabilizing ligand (SB-203580) consistently produced greater than 90% active protein on the biosensor surface. The dissociation and kinetic constants derived using this Biacore method are in excellent agreement with values determined by other methods. Additionally, we extend the method to study the thermodynamics of small molecule binding to p38alpha and derive a detailed thermodynamic reaction pathway for SB-203580. The Biacore method reported here provides an efficient way to directly and reproducibly examine dissociation constants, kinetics, and thermodynamics for small molecules binding to p38alpha and possibly other protein kinases. Immobilization in the presence of a stabilizing ligand may further represent a broadly applicable paradigm for creation of highly active biosensor surfaces.
Sigma-Aldrich
SKF-86002, ≥98% (HPLC), solid