MilliporeSigma
  • Suppressive effect of viscous dietary fiber on elevations of uric acid in serum and urine induced by dietary RNA in rats is associated with strength of viscosity.
Suppressive effect of viscous dietary fiber on elevations of uric acid in serum and urine induced by dietary RNA in rats is associated with strength of viscosity.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (2003-12-03)
Takashi Koguchi, Hisao Nakajima, Hiromi Koguchi, Masahiro Wada, Yuji Yamamoto, Satoshi Innami, Akio Maekawa, Tadahiro Tadokoro
ABSTRACT
This study was performed to clarify how dietary fiber (DF) with different viscosities would be associated with dietary RNA metabolism. Male Wistar strain rats, four weeks old, were fed diets containing a 3% (w/w) yeast RNA and a 5% (w/w) viscous DF for five days. Viscosity of DF samples used, in order of strength, were xanthan gum (XG) > guar gum (GG) > locust bean gum (LBG) > karaya gum (KG) > pectin (PE) = arabic gum (AG) > CM-cellulose (CMC) = inulin (IN). The serum uric acid concentration in the viscous DF groups significantly decreased as compared with that in the cellulose (CL) group. The urinary excretions of uric acid and allantoin in the respective groups given AG, GG, IN, KG, PE, and XG were significantly suppressed as compared with those in the CL group. The fecal RNA excretion was markedly increased in the IN, KG, PE, and XG groups in comparison to the CL group. The DF with high viscosity significantly suppressed RNA digestion by RNase A and decreased uptakes of 14C-labeled adenosine and adenosine 5'-monophosphate (5'-AMP) in rat jejunum. The results reveal that the suppressive effect of DF on elevation of serum uric acid concentration induced by dietary RNA in rats is associated with the strength of DF viscosity. The mechanism by which this is accomplished is suggested to be attributed to the inhibitions of digestion for dietary RNA and/or absorption of the hydrolyzed compounds.
Sigma-Aldrich
Karaya gum