MilliporeSigma
  • On the mechanism of the laccase-mediator system in the oxidation of lignin.
On the mechanism of the laccase-mediator system in the oxidation of lignin.
Chemistry (Weinheim an der Bergstrasse, Germany) (2003-11-13)
Claudia Crestini, Lubo Jurasek, Dimitris S Argyropoulos
ABSTRACT
In an effort to elucidate the role of phenolic and non-phenolic lignin subunits in a laccase mediator (LM) system, vanillyl alcohol was oxidized with laccase in the presence and absence of the mediator 1-hydroxybenzotriazol (HBT). Furthermore, the role of phenolic, aliphatic hydroxyl, and carboxylic acid moieties in lignin degradation was elucidated by selectively blocking them. The modified samples were then subjected to laccase and laccase-HBT treatments. On the basis of this data it was possible to establish the role of this mediator. HBT mediates the oxidation of lignin by inducing side-chain oxidation and oxygen-addition products rather than oxidative coupling reactions.
Sigma-Aldrich
4-Hydroxy-3-methoxybenzyl alcohol, 98%
Sigma-Aldrich
Vanillyl alcohol, ≥98%, FG