MilliporeSigma
  • Comparison of vasodilatory responses to nitroglycerin and its dinitrate metabolites in human veins.
Comparison of vasodilatory responses to nitroglycerin and its dinitrate metabolites in human veins.
Clinical pharmacology and therapeutics (1992-12-01)
W E Haefeli, M Gumbleton, L Z Benet, B B Hoffman, T F Blaschke
ABSTRACT
Independent of the route of nitroglycerin administration, substantial amounts of 1,2-glyceryl dinitrate (1,2-GDN) and 1,3-glyceryl dinitrate (1,3-GDN) metabolites accumulate in humans. Thus far their pharmacologic activity in comparison to nitroglycerin in humans is unknown. To compare the venodilatory potency of nitroglycerin and of 1,2-GDN and 1,3-GDN in vivo, cumulative dose-response curves were established in nine healthy volunteers by use of the dorsal hand vein compliance technique. Separated by a washout period, two of the three venodilators were infused in randomized order after preconstriction with phenylephrine. Values for maximum vasodilation were similar for all compounds: nitroglycerin, 109%; 1,2-GDN, 100%; and 1,3-GDN, 106%. The respective values for the dose rate exerting 50% of maximum vasodilation were 5.1, 43, and 60 ng/min, indicating that the dinitrates were about 10 times less potent than nitroglycerin (p < 0.001) but not significantly different from each other. The findings support the hypothesis that activity of nitroglycerin metabolites is related to the number of nitrate groups in the molecule and are in agreement with lower dinitrate potencies found in animal experiments.
Product Description
Supelco
1,3-Dinitroglycerin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®