MilliporeSigma
  • The absolute configuration of an intermediate in the asymmetric synthesis of unusual amino acids.
The absolute configuration of an intermediate in the asymmetric synthesis of unusual amino acids.
Acta crystallographica. Section C, Crystal structure communications (1992-07-15)
R Dharanipragada, M Bruck, V J Hruby
ABSTRACT
(4R)-3-[(2'R,3'R)-2'-Bromo-3'-(phenylbutanoyl)]-4-(phenylmethyl)-2 - oxazolidinone, C20H20Br-NO3, M(r) = 402.30, monoclinic, P2(1), a = 11.542 (2), b = 7.625 (1), c = 11.667 (1) A, beta = 113.97 (1) degrees, V = 938.2 (2) A3, Z = 2, Dx = 1.42 g cm-3, lambda(Mo K alpha) = 0.71073 A, mu = 21.8 cm-1, F(000) = 412, T = 296 +/- 1 K, final R = 0.028 for 2369 observed reflections. Since a D-chiral auxiliary was used the configuration at the alpha-carbon was R as expected. The two carbonyls are aligned in opposite directions to each other to overcome van der Waals repulsions.