MilliporeSigma
  • In Saccharomyces cerevisiae, cations control the fate of the energy derived from oxidative metabolism through the opening and closing of the yeast mitochondrial unselective channel.
In Saccharomyces cerevisiae, cations control the fate of the energy derived from oxidative metabolism through the opening and closing of the yeast mitochondrial unselective channel.
Journal of bioenergetics and biomembranes (2003-09-19)
Victoriano Pérez-Vázquez, Alfredo Saavedra-Molina, Salvador Uribe
ABSTRACT
The yeast mitochondrial unspecific channel (YMUC) sensitivity to inorganic (Ca2+ or Mg2+) or organic (hexyl or octyl-guanidine) cations was measured. The rate of oxygen consumption in State 3 and State 4, the transmembrane potential (deltapsi), mitochondrial swelling, and the polyethylene-glycol mediated recontraction were used to follow opening of the YMUC. Addition of 0.4 mM PO4 did not close the YMUC, although it did enhance the sensitivity to Ca2+ (I50 decreased from 50 to 0.3 mM) and Mg2+ (I50 decreased from 5 to 0.83 mM Mg2+). The Ca2+ concentration needed to close the YMUC was higher than the concentrations usually observed in the cell. Nonetheless, Mg2+, Ca2+, and PO4 exhibited additive effects. These cations did not inhibit contraction of preswollen mitochondria, suggesting that the YMUC/cation interaction was labile. Octyl-guanidine (OG-I50 7.5 microM) was the only cation which inhibited mitochondrial recontraction, probably as a result of membrane binding stabilization through its hydrophobic tail. The PO4-dependent, Ca(2+)/Mg(2+)-mediated closure of the YMUC may be a means to control the proportion of oxidative energy producing ATP or being lost as heat.