MilliporeSigma
  • Induction of human gamma globin gene expression by histone deacetylase inhibitors.
Induction of human gamma globin gene expression by histone deacetylase inhibitors.
Blood (2003-08-16)
Hua Cao, George Stamatoyannopoulos, Manfred Jung
ABSTRACT
We investigated the induction of human gamma globin gene activity by 3 classes of histone deacetylase inhibitors: amide analogues of trichostatin A, hydroxamic acid analogues of trapoxin, and scriptaid and its analogues. The screening consisted of measuring the effects of these compounds on gamma and beta human gene promoter activity by using cultures of GM979 cells stably transfected with a construct containing a gamma promoter linked to firefly luciferase and a beta promoter linked to renilla luciferase. Compounds belonging to all 3 classes induced gamma gene promoter activity in the screening assay in low micromolar concentrations. Histone deacetylase (HDAC) inhibitors increased acetylation of histone H4 and induced the expression of endogenous murine embryonic genes. They also increased the levels of gamma mRNA and the frequency of fetal hemoglobin-containing erythroblasts in erythroid burst-forming unit (BFUe) cultures from healthy adult individuals. Compounds that displayed very similar degrees of inhibition of the HDAC activity in an HDAC enzymatic assay differed strikingly on their effects on gamma gene promoter activity, raising the possibility of selectivity of HDACs that interact with the gamma globin gene chromatin.
Sigma-Aldrich
Trapoxin A, ≥98% (HPLC), from Helicoma ambiens