MilliporeSigma
SPM-927 (Schwarz Pharma).
IDrugs : the investigational drugs journal (2003-06-06)
Collin A Hovinga
ABSTRACT
Schwarz Pharma, under license from Harris FRC, is developing SPM-927, synthesized by researchers at the University of Houston, for the potential treatment of epilepsy and neuropathic pain.
Product Description
Supelco
Lacosamide solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®