MilliporeSigma
  • A sequential high-yielding large-scale solution-method for synthesis of philanthotoxin analogues.
A sequential high-yielding large-scale solution-method for synthesis of philanthotoxin analogues.
European journal of medicinal chemistry (2003-02-21)
Petrine Wellendorph, Jerzy W Jaroszewski, Steen Honoré Hansen, Henrik Franzyk
ABSTRACT
A general, improved procedure for rapid synthesis of philanthotoxin analogues, a pharmacologically important class of polyamine conjugates, is described. The solution-phase procedure is illustrated by gram-scale synthesis of philanthotoxins PhTX-343 and PhTX-12. Selectively protected polyamines are coupled to N(alpha)-Fmoc-protected amino acid pentafluorophenyl esters. After removal of the N(alpha)-Fmoc group, the amine is coupled with carboxylic acid pentafluorophenyl esters. Deprotection followed by a rapid and efficient purification by vacuum liquid chromatography on octadecylsilyl silica (RP-18 phase) gave the philanthotoxin analogues in 74-78% overall yield.