MilliporeSigma
X-ray structure of the delta-cyclodextrin complex with cycloundecanone.
Chemical communications (Cambridge, England) (2002-09-26)
Kazuaki Harata, Hiroaki Akasaka, Tomohiro Endo, Hiromasa Nagase, Haruhisa Ueda
ABSTRACT
The crystal structure of the delta-cyclodextrin complex with cycloundecanone shows a channel-type structure consisting of head-to-head dimer units that include four guest molecules.