MilliporeSigma
  • Exopolygalacturonate lyase from Thermotoga maritima: cloning, characterization and organic synthesis application.
Exopolygalacturonate lyase from Thermotoga maritima: cloning, characterization and organic synthesis application.
Carbohydrate research (2002-09-03)
Judicaël Parisot, Anahit Ghochikyan, Virginie Langlois, Vehary Sakanyan, Claude Rabiller
ABSTRACT
A new exopolygalacturonate lyase (Pel) gene of the hyperthermophilic bacterium Thermotoga maritima was cloned and overexpressed in Escherichia coli cells. A 42 kDa monomeric Pel was shown to undergo N-terminal processing by cleavage at a putative site between alanine and serine residues. The enzyme catalyzes selectively a beta-4,5 elimination at the third galacturonic unit from the reducing end of polygalacturonic acid by producing (4-deoxy-alpha-L-threo-hex-4-enopyranosyluronic acid)-(1-->4)-(alpha-D-galactopyranosyluronic acid)-(1-->4)-alpha-D-galactopyranuronic acid (3) with a 60% yield. The optimum activity of the enzyme was detected at pH 9.5 and T> or=95 degrees C. The highly thermostable enzyme constitutes a useful catalyst for a simplified synthesis of 4,5-unsaturated trigalacturonic acid 3, a trisaccharide which is extremely difficult to obtain via chemical synthesis.
Sigma-Aldrich
Trigalacturonic acid, ≥90% (HPLC)