- Pyrrolidine-5,5-trans-lactams as novel mechanism-based inhibitors of human cytomegalovirus protease. Part 3: potency and plasma stability.
Pyrrolidine-5,5-trans-lactams as novel mechanism-based inhibitors of human cytomegalovirus protease. Part 3: potency and plasma stability.
Bioorganic & medicinal chemistry letters (2002-06-18)
Alan D Borthwick, Anne M Exall, Terry M Haley, Deborah L Jackson, Andrew M Mason, Gordon G Weingarten
PMID12067545
ABSTRACT
Mechanism-based inhibitors of HCMV protease, which are stable to human plasma (> or = 20 h) and have single-figure potency in the microM range against HCMV protease, have been developed based on the dansylproline alpha-methyl pyrrolidine-5,5-trans-lactam nucleus.