MilliporeSigma
  • High-salt diet depresses acetylcholine reactivity proximal to NOS activation in cerebral arteries.
High-salt diet depresses acetylcholine reactivity proximal to NOS activation in cerebral arteries.
American journal of physiology. Heart and circulatory physiology (2002-06-14)
Francis A Sylvester, David W Stepp, Jefferson C Frisbee, Julian H Lombard
ABSTRACT
Rats were fed a low-salt (LS; 0.4% NaCl) or high-salt (HS; 4.0% NaCl) diet for 3 days, and the responses of isolated cerebral arteries to acetylcholine (ACh), the nitric oxide (NO)-dependent dilator bradykinin, and the NO donor 6-(2-hydroxy-1-methyl-2-nitrosohydrazino)-N-methyl-1-hex-anamine (NOC-9) were determined. ACh-induced vasodilation and NO release, assessed with the fluorescent NO indicator 4,5-diaminofluorescein (DAF-2) diacetate, were eliminated with the HS diet. Inhibition of cyclooxygenase, cytochrome P-450 epoxygenase, and acetylcholinesterase did not alter ACh responses. Bradykinin and NOC-9 caused a similar dilation in cerebral arteries of all groups. Arteries from animals on LS or HS diets exhibited similar levels of basal superoxide (O(2)(-)) production, assessed by dihydroethidine fluorescence, and ACh responses were unaffected by O(2)(-) scavengers. Muscarinic type 3 receptor expression was unaffected by dietary salt intake. These results indicate that 1) a HS diet attenuates ACh reactivity in cerebral arteries by inhibiting NO release, 2) this attenuation is not due to production of a cyclooxygenase-derived vasoconstrictor or elevated O(2)(-) levels, and 3) alteration(s) in ACh signaling are located upstream from NO synthase.
Sigma-Aldrich
MAHMA NONOate