MilliporeSigma
  • Efficient chemoenzymatic synthesis of globotriose and its derivatives with a recombinant alpha-(1-->4)-galactosyltransferase.
Efficient chemoenzymatic synthesis of globotriose and its derivatives with a recombinant alpha-(1-->4)-galactosyltransferase.
Carbohydrate research (2002-06-01)
Jianbo Zhang, Przemyslaw Kowal, Jianwen Fang, Peter Andreana, Peng George Wang
ABSTRACT
A truncated alpha-(1-->4)-galactosyltransferase (LgtC) gene from Neisseria meningitidis was cloned. The recombinant glycosyltransferase was expressed in Escherichia coli BL21 (DE3) strain with high specific activity (5 units/mg protein). Its acceptor specificity was carefully characterized. Then the purified enzyme was utilized in highly efficient syntheses of globotriose and a variety of alpha-(1-->4)-galactosylated derivatives as potential antibacterial agents.