MilliporeSigma
  • A (1)H NMR study of the interaction of antitumor metallocenes with glutathione.
A (1)H NMR study of the interaction of antitumor metallocenes with glutathione.
Journal of inorganic biochemistry (2001-09-22)
G Mokdsi, M M Harding
ABSTRACT
The interaction of the antitumor active metallocene dihalides Cp(2)MCl(2) (M=Ti, Nb, Mo) and 1 equiv. of glutathione was studied by (1)H NMR spectroscopy at pD 2-7 in 4 mM NaCl solutions. No interaction between glutathione and titanocene dichloride was detected at pD 2, while at pD 5-7 competitive hydrolysis of the cyclopentadienyl ligands occurred. With niobocene dichloride formation of approximately 20% of an adduct was observed at pD 2 and 5, but hydrolysis of the Cp ligands in the adduct occurred over 24 h. Molybdocene dichloride formed two stable adducts at pD 6 which were tentatively assigned as a Cp(2)Mo-glutathione chelate involving coordination of the cysteine thiol and glycine carboxylate to the metal centre, and a thiol centred 1:2 Cp(2)Mo-glutathione complex. The implications for the mechanism of antitumor action of the metallocene dihalides is discussed.