MilliporeSigma
  • Simultaneous determination of buprenorphine, norbuprenorphine, and buprenorphine-glucuronide in plasma by liquid chromatography-tandem mass spectrometry.
Simultaneous determination of buprenorphine, norbuprenorphine, and buprenorphine-glucuronide in plasma by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Biomedical sciences and applications (2001-05-08)
A Polettini, M A Huestis
ABSTRACT
For the first time, an LC-MS-MS method has been developed for the simultaneous analysis of buprenorphine (BUP), norbuprenorphine (NBUP), and buprenorphine-glucuronide (BUPG) in plasma. Analytes were isolated from plasma by C18 SPE and separated by gradient RP-LC. Electrospray ionization and MS-MS analyses were carried out using a PE-Sciex API-3000 tandem mass spectrometer. The m/z 644-->m/z 468 transition was monitored for BUPG, whereas for BUP, BUP-d4, NBUP, and NBUP-d3 it was necessary to monitor the surviving parent ions in order to achieve the required sensitivity. The method exhibited good linearity from 0.1 to 50 ng/ml (r2> or =0.998). Extraction recovery was higher than 77% for BUPG and higher than 88% for both BUP and NBUP. The LOQ was established at 0.1 ng/ml for the three analytes. The method was validated on plasma samples collected in a controlled intravenous and sublingual buprenorphine administration study. Norbuprenorphine-glucuronide was also tentatively detected in plasma by monitoring the m/z 590-->m/z 414 transition.
Sigma-Aldrich
Ammonium carbonate, puriss., meets analytical specification of NF, Ph. Franc., FCC
Sigma-Aldrich
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
Supelco
Buprenorphine glucuronide solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®