MilliporeSigma
  • Ecotoxicological effects of hexahydro-1,3,5-trinitro-1,3,5-triazine on soil microbial activities.
Ecotoxicological effects of hexahydro-1,3,5-trinitro-1,3,5-triazine on soil microbial activities.
Environmental toxicology and chemistry (2001-05-08)
P Gong, J Hawari, S Thiboutot, G Ampleman, G I Sunahara
ABSTRACT
Although hexahydro-1,3,5-trinitro-1,3,5-triazine (also called RDX or hexogen) is a potentially toxic explosive compound that persists in soil, its ecotoxicological effects on soil organisms have rarely been assessed. In this study, two uncontaminated garden soils were spiked with 10 to 12,500 mg RDX/kg dry soil. Soil microbial activities, i.e., potential nitrification, nitrogen fixation, dehydrogenase, basal respiration, and substrate-induced respiration were chosen as bioindicators and were determined after 1-, 4-, and 12-weeks of exposure. Experimental results indicate that RDX showed significant inhibition (up to 36% of control) on indigenous soil microbial communities over the period of this study. All five bioindicators responded similarly to the RDX challenge. The length of exposure also affected the microbial toxicity of RDX, with 12-week exposure exerting more significant effects than the shorter exposure periods, suggesting that soil microorganisms might become more vulnerable to RDX when exposure is extended. The estimated lowest observable adverse effect concentration of RDX was 1,235 mg/kg. No biodegradation products of RDX were detected at all three sampling times. Compared with 2,4,6-trinitrotoluene (TNT), RDX is less toxic to microbes, probably because of its resistance to biodegradation under aerobic conditions, which precludes metabolic activation of nitro groups.