MilliporeSigma
  • Agricultural "killing fields": the poisoning of Costa Rican banana workers.
Agricultural "killing fields": the poisoning of Costa Rican banana workers.
International journal of health services : planning, administration, evaluation (2000-12-08)
R Sass
ABSTRACT
The poisoning of Costa Rican banana workers by multinational corporations' excessive use of pesticides is not a local issue; it is embedded in a dominant ideology expressed by the phenomenon of globalization. This ideology seeps into every aspect of our social institutions--economic, political, and legal. The practice of this ideological perspective is evident in the industrialization of global agriculture and the shift from "developmentalism"--liberal welfarism, industrialization, and urbanization--to a dominant, undemocratic, global financial elite with "economism" and a neoliberal political agenda overriding the nation-state polis. A specific effect is to transform the agricultural workers of developing countries, such as Costa Rican banana workers, into politically superfluous flesh-and-blood human beings.
Sigma-Aldrich
1,2-Dibromo-3-chloropropane, 97%