MilliporeSigma
  • Reduction of 1,3-diphenyl-1-triazene by rat hepatic microsomes, by cecal microflora, and in rats generates the phenyl radical metabolite: nn ESR spin-trapping investigation.
Reduction of 1,3-diphenyl-1-triazene by rat hepatic microsomes, by cecal microflora, and in rats generates the phenyl radical metabolite: nn ESR spin-trapping investigation.
Chemical research in toxicology (2000-11-23)
M B Kadiiska, K S De Costa, R P Mason, J M Mathews
ABSTRACT
An ESR spin-trapping technique was used to determine whether free radical metabolites are formed as a result of the reduction of 1, 3-diphenyl-1-triazene (DPT) in vivo and in vitro by components of the cytochrome P450 (P450) mixed-function oxidase system in microsomes or by gut microflora in anaerobic cecal incubations. The ESR spectrum of the DMPO-phenyl radical adduct was detected in a microsomal incubation containing DPT, DMPO, and NADPH with the following hyperfine coupling constants: a(N) = 15.95 G and = 24.37 G. The amplitude of the spectrum from the phenyl radical adduct generated in microsomal incubations of DPT with DMPO and NADPH was not attenuated by the P450 inhibitor 1-aminobenzotriazole (ABT) or by carbon monoxide, indicating that P450 is not significantly involved in phenyl radical formation. The formation of a DMPO-phenyl radical adduct was also catalyzed by recombinant human cytochrome P450 reductase. Addition of anti-rat P450 reductase antibody led to an attenuation of the signal in incubations containing either microsomes or reductase. Low concentrations of DMPO-phenyl radical adducts were detected by ESR in the toluene extract of cecal contents containing DPT and the spin trap. In the in vivo experiments with rats treated with DPT and the spin trap DMPO, the six-line ESR signal of the DMPO-phenyl radical adduct was readily detected in bile 40-60 min after rats were treated with DPT and DMPO. The results show for the first time that the phenyl radical is formed by the reduction of DPT and may indicate a toxic potential for this chemical.