MilliporeSigma
  • Role of cytochrome P4501A1 in biotransformation of a tissue specific sarcomagen N-methyldibenzo[c,g]carbazole.
Role of cytochrome P4501A1 in biotransformation of a tissue specific sarcomagen N-methyldibenzo[c,g]carbazole.
Mutation research (2000-09-14)
A Gábelová, G Bacová, L Ruzeková, T Farkasová
ABSTRACT
7H-dibenzo[c,g]carbazole (DBC) is a potent liver and skin carcinogen, while its synthetic methyl derivative N-methyldibenzo[c,g]carbazole (MeDBC) is tissue specific sarcomagen. It is supposed that sarcomagenic activity of DBC depends on biotransformation at ring-carbon atoms, as with PAH, whereas the heterocyclic nitrogen plays an important role in liver carcinogenicity. The objective of this study was to elucidate the role of cytochrome P4501A1 in metabolic activation of sarcomagenic derivatives of DBC and to characterize the DNA damage profiles induced by DBC and MeDBC in relation to the mode of metabolic activation. The genetically engineered V79MZh1A1 cell line with stable expression of cDNA of human cytochrome P4501A1, the parental V79MZ cell line lacking any cytochrome P450 activity and human hepatocarcinoma Hep G2 cells were used as a model cells. Dose-dependent decrease in colony forming ability (CFA) was found in the V79MZh1A1 cell line after treatment of cells with DBC and MeDBC; however, no change in CFA was induced in parental V79MZ cells. These results were in a good correlation with DNA damaging effects of these two derivatives measured by the alkaline DNA unwinding (ADU) and the modified single cell gel electrophoresis (SCGE) techniques. Differences in DNA damage profiles induced by DBC and MeDBC were found in V79MZh1A1 and Hep G2 cells. These differences were probably the result of different reactive metabolite formation depending on chemical structure of the molecule and ways of biotransformation. This study showed that the cytochrome P4501A1 took part in activation of sarcomagenic DBC derivatives. Moreover, V79 cell lines with stable expression of different cytochromes P450 in combination with DNA repair endonucleases should be a useful tool for characterization of the role of individual cytochromes in metabolic activation pathways of DBC and MeDBC.
Product Description
7H-Dibenzo[c,g]carbazole, BCR®, certified reference material