- Comparison of pheromone-mediated mating disruption and conventional insecticides for management of tufted apple bud moth (Lepidoptera: Tortricidae).
Comparison of pheromone-mediated mating disruption and conventional insecticides for management of tufted apple bud moth (Lepidoptera: Tortricidae).
Large-plot studies were used to compare pheromone-mediated mating disruption and conventional insecticide applications for management of tufted apple bud moth, Platynota idaeusalis (Walker), in North Carolina in 1993 and 1994. Pheromone trap catches were reduced in mating disruption blocks, and traps placed in the lower stratum of the canopy had a higher level of trap capture reduction compared with traps placed in the upper stratum. First-generation tufted apple bud moth exposure to either pheromones for mating disruption or insecticides affected second generation pheromone trap catches in the lower and upper canopy. More second generation male moths were caught in pheromone traps placed in the upper compared with the lower canopy in blocks treated with pheromones for mating disruption during the first generation, whereas the opposite was true in blocks treated with insecticides during the first generation. Despite reduced trap catches in pheromone-treated blocks, egg mass densities were not reduced in these blocks compared with insecticide-treated blocks. Furthermore, fruit damage was not significantly different between mating disruption blocks and conventionally treated blocks in orchards with relatively low populations of tufted apple bud moth, but damage was greater in mating disruption blocks in orchards with higher moth densities.