MilliporeSigma
  • Natural and synthetic podolactones with potential use as natural herbicide models.
Natural and synthetic podolactones with potential use as natural herbicide models.
Journal of agricultural and food chemistry (2000-07-18)
F A Macías, A M Simonet, P C Pacheco, A F Barrero, E Cabrera, D Jiménez-González
ABSTRACT
A collection of 11 natural and synthetic podolactones have been tested as allelochemicals in a range between 10(-4) and 10(-9) M, and their potential use as natural herbicide models is discussed. Their effects on the germination and growth of the dicots Lactuca sativa (cv. Nigra and cv. Roman), Lepidium sativum, and Lycopersicon esculentum and the monocots Allium cepa, Hordeum vulgare, and Triticum aestivum as standard target species have been studied. An important inhibitory effect on the germination and growth of all tested species (average = 90%) was produced by compounds 9-11 at 10(-4) M. The specific structural requirements related to their activities are studied. On the basis of these results, their use as potential natural herbicide models is proposed.
Product Description
Supelco
Triasulfuron, PESTANAL®, analytical standard