MilliporeSigma
  • Chiral resolution of the enantiomers of 7-chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide using high-performance liquid chromatography on cellulose-based chiral stationary phases.
Chiral resolution of the enantiomers of 7-chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide using high-performance liquid chromatography on cellulose-based chiral stationary phases.
Journal of pharmaceutical and biomedical analysis (2000-07-18)
G Cannazza, D Braghiroli, M Baraldi, C Parenti
ABSTRACT
Analytical high-performance liquid chromatography (HPLC) methods using derivatized cellulose chiral stationary phases (CSPs) were developed for the separation of the enantiomers of 7-chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide ((+/-) IDRA21). In previous studies, (+/-) IDRA21 has been found to have an interesting inhibitory effect on the desensitization of alpha-amino-2,3-dihydro-5-methyl-3-oxo-4-isoxazolepropanoic acid (AMPA) receptor and improve cognition in animals. This compound possess one chiral carbon atom, but very little information has been reported on the stereoselectivity of his activity. Therefore resolution of the enantiomers of this compound and subsequent identification of stereospecificity in his pharmacological actions are clearly matters of interest. The resolution were made under normal- and reversed-phase conditions using a mobile phase consisting of n-hexane:2-propanol (70/30, v/v) and water:acetonitrile (60/40, v/v) respectively, and a CSP of silica-based cellulose tris-3,5-dimethyl-phenylcarbamate (Chiralcel OD and Chiracel OD-R). The enantiomeric nature of eluates was confirmed by circular dichroism (CD) spectra. A baseline separation (R(S) > 1.5) was obtained in both cases. Furthermore the isolation of optical isomers of (+/-) IDRA21 was performed using a semipreparative column packed with the same cellulose OD CSP.
Sigma-Aldrich
IDRA 21, ≥98%