MilliporeSigma
  • CYP2D6 and CYP2C19 genotypes of patients with terodiline cardiotoxicity identified through the yellow card system.
CYP2D6 and CYP2C19 genotypes of patients with terodiline cardiotoxicity identified through the yellow card system.
British journal of clinical pharmacology (2000-07-25)
G A Ford, S M Wood, A K Daly
ABSTRACT
Terodiline has concentration dependent QT prolonging effects and thus the potential for cardiotoxicity. Pharmacogenetic variation in terodiline metabolism could be responsible for cardiotoxicity. We sought to determine whether CYP2D6 (debrisoquine hydroxylase) or CYP2C19 (S-mephenytoin hydroxylase) status is a risk factor for terodiline cardiotoxicity. Using the UK Yellow Card scheme to identify patients, blood samples were obtained from eight patients who survived ventricular tachycardia or torsades de pointes suspected to be due to terodiline, for determination of CYP2D6 and CYP2C19 genotypes. Genotype prevalence was compared with that in published general population groups. One patient was a CYP2D6 poor metaboliser (CYP2D6*4 homozygous) and a second was heterozygous for CYP2D6*4, a slightly lower frequency for these genotypes compared with the general population (P = 0.31). In the case of CYP2C19, one patient was a poor metaboliser and four were heterozygous for the variant CYP2C19*2 allele, compared with general population frequencies of 2% and 23%, respectively (P = 0.035). These findings suggest that debrisoquine poor metaboliser status is not primarily responsible for terodiline cardiotoxicity. However, possession of the CYP2C19*2 allele appears to contribute to adverse cardiac reactions to terodiline. The present study demonstrates the feasibility of using spontaneous adverse drug reaction reporting schemes to determine the contribution of genotype for metabolizing enzymes to uncommon adverse drug reactions.
Sigma-Aldrich
Terodiline hydrochloride, ≥98% (HPLC), solid