MilliporeSigma
  • A preliminary study for isolation of catalytic antibodies by histidine ligand affinity chromatography as an alternative to conventional protein A/G methods.
A preliminary study for isolation of catalytic antibodies by histidine ligand affinity chromatography as an alternative to conventional protein A/G methods.
Applied biochemistry and biotechnology (2000-05-29)
E Nedonchelle, O Pitiot, M A Vijayalakshmi
ABSTRACT
Catalytic autoimmune antibodies from the sera of lupus patients were purified using histidyl-aminohexyl-Sepharose gel and compared with the antibodies purified with protein A and protein G affinity chromatography. The IgG preparations from the histidine affinity column had a much higher catalytic activity in hydrolyzing the peptide substrate Pro-Phe-Arg-methylcoumarinamide compared to the antibodies obtained by the conventional protein A/G method. This preservation of catalytic activity is attributed to the gentle buffer conditions used in the histidine ligand method that allowed the integrity of three-dimensional structure of purified catalytic antibodies. Thus, histidine affinity offer a superior method for isolating autoimmune catalytic antibodies.
Sigma-Aldrich
Pro-Phe-Arg-7-amido-4-methylcoumarin, ≥95%