MilliporeSigma
  • Antidiarrheal effects of zaldaride maleate after oral, intravenous and subcutaneous administration to rats.
Antidiarrheal effects of zaldaride maleate after oral, intravenous and subcutaneous administration to rats.
Japanese journal of pharmacology (2000-02-11)
N Aikawa, K Horikoshi, H Maeda, H Kobayashi, K Ohmori
ABSTRACT
The antidiarrheal action of zaldaride maleate (ZAL) after oral, intravenous and subcutaneous administration was examined to determine whether ZAL acts systemically or locally in the intestine of rats. Oral administration of ZAL inhibited castor oil- and 16,16-dimethyl prostaglandin E2-induced diarrhea; however, intravenous or subcutaneous administration of ZAL was ineffective. When ZAL was orally administered, the area under the plasma concentration time curve of the compound was lower than that of ZAL following intravenous or subcutaneous administration at the maximum doses studied. The antidiarrheal effect of ZAL was not dependent on its plasma concentration level. These results suggest that ZAL acts locally in the intestinal tract in rats.
Sigma-Aldrich
CGS-9343B, ≥98% (HPLC), powder