MilliporeSigma
  • Evidence for oxidative metabolism in the genotoxicity of the atmospheric reaction product 2-nitronaphthalene in human lymphoblastoid cell lines.
Evidence for oxidative metabolism in the genotoxicity of the atmospheric reaction product 2-nitronaphthalene in human lymphoblastoid cell lines.
Mutation research (1999-10-16)
J C Sasaki, J Arey, D A Eastmond, K K Parks, P T Phousongphouang, A J Grosovsky
ABSTRACT
2-Nitronaphthalene (2NN) has been identified as a mutagenic atmospheric reaction product of naphthalene in the Ames bacterial reversion assay. Recent experiments have shown this nitroarene to be genotoxic in a human lymphoblastoid cell line (MCL-5) transfected with plasmids encoding epoxide hydrolase and four cytochrome P450 monooxygenase activities. The present study investigated the genotoxicity of 2NN in two related human B-lymphoblastoid cell lines, h1A1v2 containing a single P450 isozyme (cytochrome P450 1A1) and L3 cells which are isogenic with MCL-5 cells and are distinguished only by the absence of transfected plasmids. The results indicate that 2NN-induced mutagenesis at the heterozygous thymidine kinase (tk) locus was dependent on metabolic activities provided by the transfected plasmids in MCL-5; no significant induction of mutants was observed in L3 cells studied in parallel. A similar induction of mutation was observed in h1A1v2 and MCL-5 cell lines at the tk locus and no induction was observed at the hemizygous hypoxanthine phosphoribosyl transferase (hprt) locus. The induction of mutations in h1A1v2 cells suggests that cytochrome P450 1A1 alone can activate 2NN to a mutagenic species, however, this interpretation may be confounded by differences between the h1A1v2 and MCL-5 cell lines. The observed genotoxic activity induced by 2NN prompted testing of the amino analogue, beta-naphthylamine (betaNA), to investigate potential similarities in the metabolic activation pathways of the two compounds. The negative response of betaNA in all cell lines suggests that 2NN and betaNA are not activated in these human cells by similar metabolic pathways.