MilliporeSigma
  • Effect of amlodipine in human internal mammary artery and clinical implications.
Effect of amlodipine in human internal mammary artery and clinical implications.
The Annals of thoracic surgery (2010-11-26)
Xiao-Yan Bai, Xiao-Cheng Liu, Wen-Bin Jing, Qin Yang, Xiang-Dong Tang, Guo-Wei He
ABSTRACT
Graft spasm remains challenging in CABG (coronary artery bypass grafting) surgery. We investigated the inhibitory effect of a dihydropyridine calcium antagonist amlodipine on the vasoconstriction mediated by potassium chloride (KCl), human urotensin-II (hU-II), and U46619 in human internal mammary artery (IMA) from patients undergoing CABG. Isolated IMA rings (n = 78, taken from 42 patients) were studied in organ baths in two ways: the relaxing effect of amlodipine on vasoconstrictor-induced precontraction by KCl, hU-II, and U46619 and the depressing effect of amlodipine on the contraction. Amlodipine caused full relaxation in KCl-contracted (98.0% ± 2.1%), in hU-II-contracted (98.5% ± 2.4%), and in U46619-contracted (96.3% ± 1.3%) IMA rings (n = 8) with 15.5-fold higher potency to KCl than to hU-II (effective concentration causing 50% of maximal response [EC(50)]: -8.17 ± 0.28 vs -6.98 ± 0.01 log M, p < 0.001) and 19.5-fold that to U46619 (EC(50): -8.17 ± 0.28 vs -6.88 ± 0.08 log M, p < 0.001). Pretreatment of IMA with plasma concentrations of amlodipine (-6.6 log M) significantly depressed subsequent contraction to KCl (from 20.8 ± 2.5 mN to 7.6 ± 3.0 mN, p = 0.004) and hU-II (from 14.1 ± 4.2 mN to 3.8 ± 2.0 mN, p = 0.026), but did not significantly affect the contraction to U46619. We conclude that in human IMA amlodipine has a potent inhibitory effect on the vasoconstriction mediated by a variety of vasoconstrictors. Thus, use of amlodipine in CABG patients is favored in treating and preventing graft spasm.
Sigma-Aldrich
Urotensin II human, ≥97% (HPLC), synthetic