MilliporeSigma
  • Innate immune response triggered by triacyl lipid A is dependent on phospholipid transfer protein (PLTP) gene expression.
Innate immune response triggered by triacyl lipid A is dependent on phospholipid transfer protein (PLTP) gene expression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-04-27)
Thomas Gautier, Catherine Paul, Valérie Deckert, Catherine Desrumaux, Alexis Klein, Jérôme Labbé, Naig Le Guern, Anne Athias, Serge Monier, Arlette Hammann, Ali Bettaieb, Jean-François Jeannin, Laurent Lagrost
ABSTRACT
Hexaacyl lipopolysaccharide (LPS) aggregates in aqueous media, but its partially deacylated lipid A moiety forms monomers with weaker toxicity. Because plasma phospholipid transfer protein (PLTP) transfers hexaacyl LPS, its impact on metabolism and biological activity of triacyl lipid A in mice was addressed. Triacyl lipid A bound readily to plasma high-density lipoproteins (HDLs) when active PLTP was expressed [HDL-associated lipid A after 4.5 h: 59.1+/-16.0% of total in wild-type (WT) vs. 32.5+/-10.3% in PLTP-deficient mice, P<0.05]. In the opposite to hexaacyl LPS, plasma residence time of lipid A was extended by PLTP, and proinflammatory cytokines were produced in higher amounts in WT than PLTP(-/-) mice (remaining lipid A after 8 h: 53+/-12 vs. 35+/-7%, and IL6 concentration after 4.5 h: 45.5+/-5.9 vs. 14.6+/-7.8 ng/ml, respectively; P<0.05 in all cases). After 1 wk, onset of B16-induced melanoma was observed in only 30% of lipid A-treated WT mice, whereas >80% of the untreated WT, untreated PLTP-deficient, or lipid A-treated PLTP-deficient animals bore tumors (P<0.05 in all cases). It is concluded that PLTP is essential in mediating the association of triacyl lipid A with lipoproteins, leading to extension of its residence time and to magnification of its proinflammatory and anticancer properties.
Sigma-Aldrich
PLTP Activity Assay Kit, Supplied by Roar Biomedical, Inc.