MilliporeSigma
  • Liver X receptor-mediated induction of cholesteryl ester transfer protein expression is selectively impaired in inflammatory macrophages.
Liver X receptor-mediated induction of cholesteryl ester transfer protein expression is selectively impaired in inflammatory macrophages.
Arteriosclerosis, thrombosis, and vascular biology (2009-08-15)
Daniela Lakomy, Cédric Rébé, Anne-Laure Sberna, David Masson, Thomas Gautier, Angélique Chevriaux, Magalie Raveneau, Nicolas Ogier, Anh Thoai Nguyen, Philippe Gambert, Jacques Grober, Bernard Bonnotte, Eric Solary, Laurent Lagrost
ABSTRACT
Cholesteryl ester transfer protein (CETP) is a target gene for the liver X receptor (LXR). The aim of this study was to further explore this regulation in the monocyte-macrophage lineage and its modulation by lipid loading and inflammation, which are key steps in the process of atherogenesis. Exposure of bone marrow-derived macrophages from human CETP transgenic mice to the T0901317 LXR agonist increased CETP, PLTP, and ABCA1 mRNA levels. T0901317 also markedly increased CETP mRNA levels and CETP production in human differentiated macrophages, whereas it had no effect on CETP expression in human peripheral blood monocytes. In inflammatory mouse and human macrophages, LXR-mediated CETP gene upregulation was inhibited, even though ABCA1, ABCG1, and SREBP1c inductions were maintained. The inhibition of CETP gene response to LXR agonists in inflammatory cells was independent of lipid loading (ie, oxidized LDL increased CETP production in noninflammatory macrophages with a synergistic effect of synthetic LXR agonists). LXR-mediated induction of human CETP expression is switched on during monocyte-to-macrophage differentiation, is magnified by lipid loading, and is selectively lost in inflammatory macrophages, which suggests that inflammatory cells may not increase the circulating CETP pool on LXR agonist treatment.
Sigma-Aldrich
CETP Activity Assay Kit, Supplied by Roar Biomedical, Inc.
Sigma-Aldrich
PLTP Activity Assay Kit, Supplied by Roar Biomedical, Inc.