MilliporeSigma
  • Arginine vasopressin enhances pHi regulation in the presence of HCO3- by stimulating three acid-base transport systems.
Arginine vasopressin enhances pHi regulation in the presence of HCO3- by stimulating three acid-base transport systems.
Nature (1989-02-16)
M B Ganz, G Boyarsky, R B Sterzel, W F Boron
ABSTRACT
Growth factors raise intracellular pH (pHi) by stimulating Na+/H+ exchange in the absence of HCO3-. In mutant cells that lack the Na+/H+ exchange activity, this alkalinization does not occur, and the cells do not proliferate without artificial elevation of pHi. It has therefore been widely suggested that an early pHi increase is a necessary signal for mitogenesis. In the presence of HCO3- however, growth factors fail to raise pHi in A431 cells, renal mesangial cells and 3T3 fibroblasts. In mesangial cells, arginine vasopressin (AVP) raises pHi in the absence of HCO3-, but lowers it when HCO3- is present; growth is stimulated under both conditions. We report here that, in the presence of HCO3-, AVP stimulates two potent HCO3- transporters, as well as the Na+/H+ exchanger. These are the Na+-dependent and Na+-independent Cl-/HCO3- exchangers. Our results indicate that AVP causes acidification in the presence of HCO3- because, at the resting pHi, it stimulates Na+-independent Cl-/HCO3- exchange (which lowers pHi) more than it stimulates the sum of Na+/H+ exchange and Na+-dependent Cl-/HCO3- exchange (both of which raise pHi). The stimulation of three acid-base transporters by the growth factor AVP greatly enhances the ability of the cell to regulate pHi.
Sigma-Aldrich
[deamino-Cys1, Val4, D-Arg8]-Vasopressin, ≥95%
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-me-Tyr2, Arg8]-Vasopressin, ≥97% (HPLC)
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl
Sigma-Aldrich
[deamino-Cys1, D-Arg8]-Vasopressin acetate salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-Et-Tyr2, Val4, Arg8]-Vasopressin, ≥97% (HPLC)