MilliporeSigma
  • Regulation of sympathetic nerve activity by L-carnosine in mammalian white adipose tissue.
Regulation of sympathetic nerve activity by L-carnosine in mammalian white adipose tissue.
Neuroscience letters (2008-07-05)
Jiao Shen, Jia-Fei Yao, Mamoru Tanida, Katsuya Nagai
ABSTRACT
Previously, we showed that l-carnosine, a bioactive dipeptide, influences the sympathetic nerve activity innervating kidney and brown adipose tissue. Because the autonomic nervous system plays an important role in the regulation of lipid metabolism, we investigated the in vivo effects of L-carnosine on the sympathetic nerve activity innervating white adipose tissue (SNA-WAT) and lipolysis. We found that intraperitoneal (ip) administration of L-carnosine at doses of 100 ng/rat and 10 microg/rat elevated and suppressed SNA-WAT, respectively. The effect of lower dose of L-carnosine (100 ng/rat) was eliminated by pretreatment with diphenhydramine hydrochloride, a histamine H(1) receptor antagonist. In contrast, the effect of higher dose of L-carnosine (10 microg/rat) was suppressed by thioperamide maleate salt, a histamine H(3) receptor antagonist. Moreover, ip administration of 100 ng and 10 microg of L-carnosine increased and decreased the levels of plasma free fatty acids (FFAs), respectively. The changes of plasma FFAs resulting from the exposure to 100 ng and 10 microg of L-carnosine were diminished by the beta-adrenergic receptor blocker propranolol hydrochloride and the muscarinic receptor blocker atropine sulfate, respectively; and eliminated by the corresponding histamine receptor antagonists, which eliminated the changes in SNA-WAT. Our results suggest that low doses of L-carnosine may regulate the lipolytic processes in adipose tissue through facilitation of the sympathetic nervous system, which is driven by histamine neurons through the H(1) receptor, and that the beta(3)-receptor may be involved in this enhanced lipolytic response. High doses of L-carnosine, on the other hand, may lower lipolysis by suppressing sympathetic nerve activity via the H(3) receptor, and the muscarinic receptor may be related to this response.
Sigma-Aldrich
Thioperamide maleate salt, solid