MilliporeSigma
  • The kynurenine 3-hydroxylase inhibitor Ro 61-8048 improves dystonia in a genetic model of paroxysmal dyskinesia.
The kynurenine 3-hydroxylase inhibitor Ro 61-8048 improves dystonia in a genetic model of paroxysmal dyskinesia.
European journal of pharmacology (2003-10-14)
Angelika Richter, Melanie Hamann
ABSTRACT
The effects of the novel kynurenine 3-hydroxylase inhibitor 3,4-dimethoxy-N-[4-(3-nitrophenyl)thiazol-2-yl]benzenesulfonamide (Ro 61-8048) on severity of dystonia were examined in dt(sz) mutant hamsters, an animal model of paroxysmal dystonia, in which stress precipitates dystonic episodes. Ro 61-8048 (50, 100 and 150 mg/kg i.p.) significantly reduced the severity of dystonia in dt(sz) hamsters without leading to marked central side effects. Determinations of kynurenic acid concentrations in brain homogenates demonstrated that Ro 61-8048 (100 mg/kg i.p.) provoked a two- to threefold increase of the endogeneous broad spectrum glutamate receptor antagonist kynurenic acid in the striatum, cerebellum and brainstem of mutant hamsters. The antidystonic efficacy of Ro 61-8048 at well-tolerated doses suggests that kynurenine 3-hydroxylase inhibitors should be considered as new therapeutic candidates for the treatment of dyskinesias.
Sigma-Aldrich
Ro 61-8048, ≥98% (HPLC)