MilliporeSigma
  • Studies on the mechanism of superoxide release from human neutrophils stimulated with arachidonate.
Studies on the mechanism of superoxide release from human neutrophils stimulated with arachidonate.
The Journal of biological chemistry (1984-10-10)
J T Curnutte, J A Badwey, J M Robinson, M J Karnovsky, M L Karnovsky
ABSTRACT
cis-Unsaturated fatty acids stimulate release of superoxide (O-2) by human neutrophils (Badwey, J. A., Curnutte, J. T., Robinson, J. M., Berde, C. B., Karnovsky, M. J., and Karnovsky, M. L. (1984) J. Biol. Chem. 259, 7870-7877). The rate of O-2 release due to arachidonate (105 +/- 24 S.D., nmol of O-2/min/10(7) cells) was comparable to optimal values obtained with other stimuli. Antagonists of calcium-binding proteins (i.e. phenothiazines, naphthalene sulfonamides) inhibited the release of O-2 in a fashion compatible with the involvement of calmodulin in these phenomena. Synthetic substrates for and an inhibitor of chymotrypsin-like proteases (e.g. N-benzoyl-L-tyrosine ethyl ester, L-1-tosylamido-2-phenylethyl chloromethyl ketone) also blocked O-2 release. Antagonists of calcium-binding proteins and of proteases were effective in this context with neutrophils stimulated with a variety of agents. The implications of these data for recent reports concerning the mechanism of action of cis-unsaturated fatty acids on phagocytes is discussed.
Sigma-Aldrich
N-Benzoyl-L-tyrosine ethyl ester