- [Detection of disordered immune responses to the chronic effects of vinyl chloride in the organism (clinical and experimental study)].
[Detection of disordered immune responses to the chronic effects of vinyl chloride in the organism (clinical and experimental study)].
Meditsina truda i promyshlennaia ekologiia (2013-06-22)
G M Bodienkova, R Iu Alekseev, E V Boklazhenko
PMID23785824
ABSTRACT
This paper considers the features of disorders of immunity in chronic exposure to vinyl chloride at work and in experimental animals. It is shown that one of the earliest and most sensitive indicators of the formation of the resistance of the organism employed in the manufacture of vinyl chloride, are: IgM, IL-1beta, TNF-alpha, autoantibodies to neurospecific proteins (S-100, Vd-Ca channels and DNA).