MilliporeSigma
  • Cost-effectiveness of statins for primary cardiovascular prevention in chronic kidney disease.
Cost-effectiveness of statins for primary cardiovascular prevention in chronic kidney disease.
Journal of the American College of Cardiology (2013-03-19)
Kevin F Erickson, Sohan Japa, Douglas K Owens, Glenn M Chertow, Alan M Garber, Jeremy D Goldhaber-Fiebert
ABSTRACT
The authors sought to evaluate the cost-effectiveness of statins for primary prevention of myocardial infarction (MI) and stroke in patients with chronic kidney disease (CKD). Patients with CKD have an elevated risk of MI and stroke. Although HMG Co-A reductase inhibitors (“statins”) may prevent cardiovascular events in patients with non–dialysis-requiring CKD, adverse drug effects and competing risks could materially influence net effects and clinical decision-making. We developed a decision-analytic model of CKD and cardiovascular disease (CVD) to determine the cost-effectiveness of low-cost generic statins for primary CVD prevention in men and women with hypertension and mild-to-moderate CKD. Outcomes included MI and stroke rates, discounted quality-adjusted life years (QALYs) and lifetime costs (2010 USD), and incremental cost-effectiveness ratios. For 65-year-old men with moderate hypertension and mild-to-moderate CKD, statins reduced the combined rate of MI and stroke, yielded 0.10 QALYs, and increased costs by $1,800 ($18,000 per QALY gained). For patients with lower baseline cardiovascular risks, health and economic benefits were smaller; for 65-year-old women, statins yielded 0.06 QALYs and increased costs by $1,900 ($33,400 per QALY gained). Results were sensitive to rates of rhabdomyolysis and drug costs. Statins are less cost-effective when obtained at average retail prices, particularly in patients at lower CVD risk. Although statins reduce absolute CVD risk in patients with CKD, the increased risk of rhabdomyolysis, and competing risks associated with progressive CKD, partly offset these gains. Low-cost generic statins appear cost-effective for primary prevention of CVD in patients with mild-to-moderate CKD and hypertension.
Sigma-Aldrich
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.