MilliporeSigma
Application drying time for oxybutynin chloride topical gel.
Urologic nursing (2013-03-12)
Diane K Newman, Scott M Olsen, Naomi V Dahl
ABSTRACT
This study determined the median drying time for oxybutynin chloride topical gel (OTG) approved for treatment of overactive bladder was 3.1 minutes, Most participants reported positive perceptions of OTG attributes.
Sigma-Aldrich
Oxybutynin chloride, meets EP, USP testing specifications
Sigma-Aldrich
Oxybutynin chloride, ≥98% (TLC), powder