MilliporeSigma
Flupirtine, a re-discovered drug, revisited.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2013-01-17)
Istvan Szelenyi
ABSTRACT
Flupirtine was developed long before K(V)7 (KCNQ) channels were known. However, it was clear from the beginning that flupirtine is neither an opioid nor a nonsteroidal anti-inflammatory analgesic. Its unique muscle relaxing activity was discovered by serendipity. In the meantime, broad and intensive research has resulted in a partial clarification of its mode of action. Flupirtine is the first therapeutically used K(V)7 channel activator with additional GABA(A)ergic mechanisms and thus the first representative of a novel class of analgesics. The presently accepted main mode of its action, potassium K(V)7 (KCNQ) channel activation, opens a series of further therapeutic possibilities. One of them has now been realized: its back-up compound, the bioisostere retigabine, has been approved for the treatment of epilepsy.
Sigma-Aldrich
Flupirtine maleate salt, ≥98% (HPLC)