MilliporeSigma
  • Iron telluride nanorods-based system for the detection of total mercury in blood.
Iron telluride nanorods-based system for the detection of total mercury in blood.
Journal of hazardous materials (2012-11-21)
Prathik Roy, Zong-Hong Lin, Chi-Te Liang, Huan-Tsung Chang
ABSTRACT
We have developed a simple, colorimetric iron telluride (FeTe) nanorods (NRs) based system for the detection of mercury, mainly based on the cation exchange reaction between FeTe NRs and Hg(2+). FeTe NRs (length, 105 ± 21 nm) react with Hg(2+) to form HgTe NRs (length, 112 ± 26 nm) and consequently release Fe(2+) ions that catalyzes the oxidation between a peroxidase substrate 2,2'-azino-bis(3-ethylbenzo-thiazoline-6-sulfonic acid) diammonium salt (ABTS) and H(2)O(2). The concentration of Fe(2+) and thereby Hg(2+) can be determined by measuring the absorbance of the ABTS oxidized product at 418 nm. This approach allows the detection of Hg(2+), with a limit of detection of 1.31 nM at a signal-to-noise ratio 3 and a linear range 5-100 nM (R(2)=0.99). The low-cost, simple, sensitive, and reproducible assay has been validated for the detection of Hg(2+) in a blood sample (SRM 955c), with the result being in good agreement with that provided by National Institute of Standards and Technology.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)