MilliporeSigma
  • Protective effect of diltiazem, a L-type calcium channel antagonist, on lysophosphatidylcholine-enhanced hydroxyl radical generation by MPP(+) in rat striatum.
Protective effect of diltiazem, a L-type calcium channel antagonist, on lysophosphatidylcholine-enhanced hydroxyl radical generation by MPP(+) in rat striatum.
Clinical biochemistry (2012-09-25)
Toshio Obata, Masahiro Miyashita
ABSTRACT
The interaction between the PKC and oxygen free radicals. The present study examined the effects of lysophosphatidylcholine (LPC), an endogenous amphophilic lipid metabolite, on 1-methyl-4-phenylpyridinium ion (MPP(+))-induced hydroxyl radical (()OH) generation in rat striatum. LPC (50 μM) increased ()OH formation by MPP(+). Further, the effect of diltiazem, a L-type Ca(2+) channel antagonist, on MPP(+) (5mM)-induced ()OH generation was studied. Diltiazem (100 μM) significantly suppressed LPC and MPP(+)-induced ()OH generation. The results indicate ()OH generation, and diltiazem may have preventive effect on MPP(+)-induced ()OH generation. These results demonstrated that diltiazem suppressed LPC-induced generation of free radical through blockage of Ca(2+) channel in the rat striatum.
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Diltiazem hydrochloride solution, 1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®