MilliporeSigma
  • Evaluating insulin secretagogues in a humanized mouse model with functional human islets.
Evaluating insulin secretagogues in a humanized mouse model with functional human islets.
Metabolism: clinical and experimental (2012-09-18)
Jian Luo, Kathy Nguyen, Michael Chen, Than Tran, Jianqiang Hao, Bole Tian, Ingrid C Rulifson, Ying Zhang, Lei Tian, Yu Zhang, Edwin Lopez, Daniel C-H Lin, Yingcai Wang, Zhihua Ma, Jonathan Houze, Zhiguang Guo
ABSTRACT
To develop a rapid, easy and clinically relevant in vivo model to evaluate novel insulin secretagogues on human islets, we investigated the effect of insulin secretagogues on functional human islets in a humanized mouse model. Human islets were transplanted under the kidney capsule of streptozotocin (STZ)-induced diabetic mice with immunodeficiency. Human islet graft function was monitored by measuring non-fasting blood glucose levels. After diabetes was reversed, human islet transplanted mice were characterized physiologically by oral glucose tolerance and pharmacologically with clinically proven insulin secretagogues, glucagon-like peptide-1 (GLP-1), exenatide, glyburide, nateglinide and sitagliptin. Additionally, G protein-coupled receptor 40 (GPR40) agonists were evaluated in this model. Long-term human islet graft survival could be achieved in immunodeficient mice. Oral glucose challenge in human islet transplanted mice resulted in an immediate incremental increase of plasma human C-peptide, while the plasma mouse C-peptide was undetectable. Treatments with GLP-1, exenatide, glyburide, nateglinide and sitagliptin effectively increased plasma human C-peptide levels and improved postprandial glucose concentrations. GPR40 agonists also stimulated human C-peptide secretion and significantly improved postprandial glucose in the human islet transplanted mice. Our studies indicate that a humanized mouse model with human islet grafts could mimic the in vivo characteristics of human islets and could be a powerful tool for the evaluation of novel insulin secretagogues or other therapeutic agents that directly and/or indirectly target human β cells.
Sigma-Aldrich
Nateglinide, ≥98% (HPLC), solid
N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, European Pharmacopoeia (EP) Reference Standard