MilliporeSigma
  • Hyperinsulinemia euglycemia therapy for calcium channel blocker overdose: a case report.
Hyperinsulinemia euglycemia therapy for calcium channel blocker overdose: a case report.
Texas Heart Institute journal (2012-09-06)
Anushree Agarwal, Siegfried W Yu, Abdul Rehman, Joseph Q Henkle
ABSTRACT
We report the case of a patient with calcium channel blocker toxicity who was treated successfully with hyperinsulinemia euglycemia therapy, without prior use of vasopressors. The patient was a 60-year-old man with schizoaffective disorder who presented with severe hemodynamic compromise after an intentional overdose of 5,400 mg of extended-release diltiazem. He had been admitted to the hospital twice before for attempted suicide with diltiazem and nifedipine, respectively. During the previous admissions, conventional treatments were used, and complications included hemodynamic compromise, ischemic bowel requiring ileostomy, and a prolonged hospital stay. During the current admission, the patient's clinical condition failed to improve after treatment with charcoal, fluid resuscitation, calcium, and glucagon. Eight hours after admission, hyperinsulinemia euglycemia therapy was initiated; 3 hours later, the patient's hemodynamic status showed sustained improvement. His bradycardia and hypotension resolved without cardiac pacing or vasopressors. Hyperinsulinemia euglycemia therapy is a potentially life-saving treatment for calcium channel blocker toxicity. We suggest that such therapy should be considered early, in conjunction with conventional therapy, for the treatment of calcium channel blocker overdose in patients not responding to initial treatment.
Product Description
Supelco
Diltiazem hydrochloride solution, 1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard