MilliporeSigma
  • (1)H NMR-based metabonomic profiling of rat serum and urine to characterize the subacute effects of carbamate insecticide propoxur.
(1)H NMR-based metabonomic profiling of rat serum and urine to characterize the subacute effects of carbamate insecticide propoxur.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2012-07-12)
Yu-Jie Liang, Hui-Ping Wang, Ding-Xin Long, Yi-Jun Wu
ABSTRACT
Carbamate insecticide propoxur is widely used in agriculture and public health programs. To prevent adverse health effects arising from exposure to this insecticide, sensitive methods for detection of early stage organismal changes are necessary. We present here an integrative metabonomic approach to investigate toxic effects of pesticide in experimental animals. Results showed that propoxur even at low dose levels can induce oxidative stress, impair liver function, enhance ketogenesis and fatty acid β-oxidation, and increase glycolysis, which contribute to the hepatotoxocity. These findings highlight the applicability of (1)H NMR spectroscopy and multivariate statistics in elucidating the toxic effects of propoxur.
Product Description
Supelco
Propoxur, PESTANAL®, analytical standard